The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014825.2 | NP_055640.2 | 31 - 32 | 5211 | 58 |
GenBank mRNA | AB011111.2 | - | 26 - 27 | 4445 | 58 |
AF231919.1 | - | 31 - 32 | 5049 | 58 |