Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCCHC14
Entrez Gene ID
23174
Gene Name
zinc finger CCHC-type containing 14
Gene Aliases
BDG-29, BDG29
Chromosome Location
Chr.16: 87406246 - 87492948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015144.2 NP_055959.1 1 - 2 185 92
XM_005255858.3 XP_005255915.2 1 - 2 376 92
XM_011522964.2 XP_011521266.1 1 - 2 1279 92
GenBank mRNA AB011151.2 - 1 - 2 43 92
AB030243.1 - 1 - 2 193 92
AK289682.1 - 1 - 2 376 92
BC101478.1 - 1 - 2 192 92

View Full Product Details