The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014010.4 | NP_054729.3 | 12 - 13 | 2348 | 90 |
GenBank mRNA | AB014534.1 | - | 12 - 13 | 2348 | 90 |
AF116574.1 | - | 10 - 11 | 1569 | 90 | |
AK127350.1 | - | 11 - 12 | 1569 | 90 | |
AK315025.1 | - | 10 - 11 | 1412 | 90 | |
BC018759.2 | - | 6 - 7 | 797 | 90 | |
BC146756.1 | - | 12 - 13 | 2348 | 90 | |
BX649176.1 | - | 12 - 13 | 1683 | 90 |