Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPHP4
Entrez Gene ID
261734
Gene Name
nephronophthisis 4
Gene Aliases
POC10, SLSN4
Chromosome Location
Chr.1: 5862808 - 5992473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291593.1 NP_001278522.1 15 - 16 2441 62
NM_001291594.1 NP_001278523.1 14 - 15 2264 62
NM_015102.4 NP_055917.1 18 - 19 2751 62
NR_111987.1 18 - 19 2748 62
XM_006710563.3 XP_006710626.1 18 - 19 2597 62
XM_011541213.1 XP_011539515.1 17 - 18 2518 62
XM_011541214.1 XP_011539516.1 17 - 18 2521 62
XM_011541215.1 XP_011539517.1 16 - 17 2410 62
XM_011541216.2 XP_011539518.1 19 - 20 3225 62
XM_011541217.2 XP_011539519.1 18 - 19 3089 62
XM_011541218.2 XP_011539520.1 19 - 20 2758 62
XM_017000996.1 XP_016856485.1 17 - 18 2518 62
XM_017000997.1 XP_016856486.1 18 - 19 3020 62
XM_017000998.1 XP_016856487.1 17 - 18 2521 62
XM_017000999.1 XP_016856488.1 18 - 19 2516 62
XM_017001000.1 XP_016856489.1 17 - 18 2349 62
XM_017001001.1 XP_016856490.1 12 - 13 1788 62
XM_017001002.1 XP_016856491.1 17 - 18 2521 62
XM_017001003.1 XP_016856492.1 10 - 11 1579 62
GenBank mRNA AB014573.1 - 13 - 14 1851 62
AF537130.1 - 18 - 19 2749 62
AK124074.1 - 14 - 15 2217 62
AK314326.1 - 15 - 16 2415 62
AY118228.1 - 18 - 19 2686 62
BC040520.1 - 18 - 19 2749 62

View Full Product Details