The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005255867.1 | XP_005255924.1 | 14 - 15 | 1728 | 68 |
XM_005255868.1 | XP_005255925.1 | 15 - 16 | 1740 | 68 | |
XM_011522970.1 | XP_011521272.1 | 15 - 16 | 1740 | 68 | |
XM_011522971.2 | XP_011521273.1 | 15 - 16 | 1740 | 68 | |
XM_011522973.2 | XP_011521275.1 | 15 - 16 | 1740 | 68 | |
XM_017023094.1 | XP_016878583.1 | 15 - 16 | 1740 | 68 | |
XM_017023095.1 | XP_016878584.1 | 15 - 16 | 1740 | 68 | |
XM_017023096.1 | XP_016878585.1 | 15 - 16 | 1740 | 68 | |
XM_017023097.1 | XP_016878586.1 | 15 - 16 | 1740 | 68 | |
GenBank mRNA | AB023222.1 | - | 10 - 11 | 1157 | 68 |
AK096333.1 | - | 14 - 15 | 1739 | 68 | |
AK304790.1 | - | 14 - 15 | 1790 | 68 | |
AK307929.1 | - | 14 - 15 | 1748 | 68 | |
AK308390.1 | - | 14 - 15 | 1739 | 68 | |
BC136433.1 | - | 14 - 15 | 1733 | 68 |