Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOCK9
Entrez Gene ID
23348
Gene Name
dedicator of cytokinesis 9
Gene Aliases
ZIZ1, ZIZIMIN1
Chromosome Location
Chr.13: 98793487 - 99086406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130048.1 NP_001123520.1 52 - 53 5940 60
NM_001318849.1 NP_001305778.1 51 - 52 5754 60
NM_015296.2 NP_056111.1 52 - 53 5823 60
XM_005254034.3 XP_005254091.1 49 - 50 5770 60
XM_005254035.3 XP_005254092.1 49 - 50 5677 60
XM_006719922.1 XP_006719985.1 52 - 53 6042 60
XM_006719924.2 XP_006719987.1 52 - 53 5881 60
XM_006719925.1 XP_006719988.1 52 - 53 6009 60
XM_006719931.3 XP_006719994.1 52 - 53 5768 60
XM_006719932.1 XP_006719995.1 52 - 53 5940 60
XM_006719936.1 XP_006719999.1 51 - 52 5904 60
XM_006719937.3 XP_006720000.1 51 - 52 5699 60
XM_017020503.1 XP_016875992.1 52 - 53 5882 60
XM_017020504.1 XP_016875993.1 50 - 51 5840 60
XM_017020505.1 XP_016875994.1 51 - 52 5813 60
XM_017020506.1 XP_016875995.1 52 - 53 5789 60
XM_017020507.1 XP_016875996.1 49 - 50 5771 60
XM_017020508.1 XP_016875997.1 50 - 51 5747 60
XM_017020509.1 XP_016875998.1 51 - 52 6003 60
XM_017020510.1 XP_016875999.1 51 - 52 5720 60
XM_017020511.1 XP_016876000.1 49 - 50 5702 60
XM_017020512.1 XP_016876001.1 50 - 51 5967 60
XM_017020513.1 XP_016876002.1 49 - 50 5678 60
XM_017020514.1 XP_016876003.1 50 - 51 5934 60
XM_017020515.1 XP_016876004.1 52 - 53 5916 60
XM_017020516.1 XP_016876005.1 49 - 50 5898 60
XM_017020517.1 XP_016876006.1 50 - 51 5813 60
XM_017020518.1 XP_016876007.1 52 - 53 5912 60
XM_017020519.1 XP_016876008.1 50 - 51 5841 60
XM_017020520.1 XP_016876009.1 49 - 50 5805 60
XM_017020521.1 XP_016876010.1 51 - 52 5904 60
GenBank mRNA AB028981.2 - 52 - 53 5846 60
AF527605.1 - 52 - 53 5823 60
AK127215.1 - 13 - 14 2837 60
AK294573.1 - 15 - 16 1754 60
AK294705.1 - 24 - 25 2594 60
AK294770.1 - 13 - 14 1927 60
AK300298.1 - 14 - 15 1783 60
AK300331.1 - 16 - 17 1938 60
BC043506.1 - 9 - 10 1077 60
BC053620.1 - 9 - 10 1177 60
BC136274.1 - 51 - 52 5734 60
BC171753.1 - 51 - 52 5734 60
BX648707.1 - 12 - 13 3594 60
CR749658.1 - 13 - 14 2764 60
CR936820.1 - 48 - 49 10141 60

View Full Product Details