The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020546.2 | NP_065433.2 | 6 - 7 | 1043 | 101 |
XM_011513942.2 | XP_011512244.1 | 6 - 7 | 990 | 101 | |
GenBank mRNA | AB028983.1 | - | 3 - 4 | 364 | 101 |
AF410885.1 | - | 6 - 7 | 1002 | 101 | |
AK123013.1 | - | 5 - 6 | 944 | 101 | |
AK127036.1 | - | 6 - 7 | 991 | 101 | |
AK294555.1 | - | 3 - 4 | 499 | 101 | |
AK295531.1 | - | 3 - 4 | 383 | 101 | |
AK299775.1 | - | 5 - 6 | 523 | 101 | |
AK304041.1 | - | 6 - 7 | 878 | 101 | |
BC111743.1 | - | 6 - 7 | 981 | 101 |