The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080467.2 | NP_001073936.1 | 18 - 19 | 2486 | 63 |
GenBank mRNA | AB032945.2 | - | 18 - 19 | 2213 | 63 |
AB290160.1 | - | 18 - 19 | 2303 | 63 |