The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318052.1 | NP_001304981.1 | 2 - 3 | 1412 | 78 |
NM_001318053.1 | NP_001304982.1 | 3 - 4 | 1322 | 78 | |
NM_015689.4 | NP_056504.3 | 3 - 4 | 1514 | 78 | |
NR_134477.1 | 1 - 2 | 1139 | 78 | ||
XM_005249977.3 | XP_005250034.1 | 3 - 4 | 2045 | 78 | |
XM_005249979.4 | XP_005250036.1 | 3 - 4 | 1298 | 78 | |
XM_011516052.2 | XP_011514354.1 | 3 - 4 | 1392 | 78 | |
XM_011516053.2 | XP_011514355.1 | 2 - 3 | 1496 | 78 | |
XM_011516054.2 | XP_011514356.1 | 3 - 4 | 1300 | 78 | |
XM_017011989.1 | XP_016867478.1 | 2 - 3 | 1498 | 78 | |
GenBank mRNA | AB033103.2 | - | 2 - 3 | 1412 | 78 |
AK290056.1 | - | 3 - 4 | 1283 | 78 | |
AK297959.1 | - | 2 - 3 | 1181 | 78 | |
BC115004.1 | - | 3 - 4 | 1322 | 78 | |
BC132875.1 | - | 1 - 2 | 1109 | 78 |