The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170629.1 | NP_001164100.1 | 30 - 31 | 5662 | 78 |
NM_020920.3 | NP_065971.2 | 31 - 32 | 4907 | 78 | |
GenBank mRNA | AB046784.3 | - | 30 - 31 | 5150 | 78 |
AK056639.1 | - | 11 - 12 | 1529 | 78 | |
AK131077.1 | - | 16 - 17 | 2486 | 78 | |
BC098452.1 | - | 5 - 6 | 578 | 78 | |
CR749315.1 | - | 31 - 32 | 4852 | 78 | |
U00930.1 | - | - | 1678 | 78 |