The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142640.1 | NP_001136112.1 | 21 - 22 | 5426 | 58 |
NM_018996.3 | NP_061869.2 | 20 - 21 | 5318 | 58 | |
XM_006721996.3 | XP_006722059.3 | 22 - 23 | 11087 | 58 | |
XM_006721997.3 | XP_006722060.3 | 21 - 22 | 10937 | 58 | |
XM_006722000.3 | XP_006722063.3 | 21 - 22 | 10961 | 58 | |
XM_011525088.2 | XP_011523390.1 | 19 - 20 | 5124 | 58 | |
XM_017024906.1 | XP_016880395.1 | 22 - 23 | 11078 | 58 | |
XM_017024907.1 | XP_016880396.1 | 21 - 22 | 10970 | 58 | |
XM_017024908.1 | XP_016880397.1 | 21 - 22 | 10862 | 58 | |
GenBank mRNA | AB046802.2 | - | 17 - 18 | 4868 | 58 |
AK056421.1 | - | 10 - 11 | 1401 | 58 | |
AK092762.1 | - | 5 - 6 | 671 | 58 | |
AL834429.1 | - | 17 - 18 | 2840 | 58 | |
BC045631.1 | - | 18 - 19 | 2647 | 58 |