The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020959.2 | NP_066010.1 | 1 - 2 | 265 | 91 |
XM_017027048.1 | XP_016882537.1 | 1 - 2 | 270 | 91 | |
GenBank mRNA | AB046843.2 | - | 1 - 2 | 265 | 91 |