Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTYH3
Entrez Gene ID
80727
Gene Name
tweety family member 3
Gene Aliases
-
Chromosome Location
Chr.7: 2631969 - 2664802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025250.2 NP_079526.1 4 - 5 812 56
XM_011515535.2 XP_011513837.1 4 - 5 780 56
GenBank mRNA AB051478.2 - 4 - 5 807 56
AB162931.1 - 4 - 5 637 56
BC131824.1 - 4 - 5 683 56
BC152447.1 - 4 - 5 807 56

View Full Product Details