The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003502.3 | NP_003493.1 | 8 - 9 | 2580 | 52 |
XM_011522682.2 | XP_011520984.1 | 8 - 9 | 2606 | 52 | |
XM_011522684.2 | XP_011520986.1 | 8 - 9 | 2456 | 52 | |
XM_011522686.1 | XP_011520988.1 | 7 - 8 | 1363 | 52 | |
XM_017023743.1 | XP_016879232.1 | 8 - 9 | 2591 | 52 | |
XM_017023745.1 | XP_016879234.1 | 7 - 8 | 1631 | 52 | |
XM_017023746.1 | XP_016879235.1 | 7 - 8 | 2207 | 52 | |
GenBank mRNA | AF009674.1 | - | 8 - 9 | 2306 | 52 |
BC017447.1 | - | 3 - 4 | 782 | 52 | |
BC035872.1 | - | 7 - 8 | 1621 | 52 | |
KF615847.1 | - | 8 - 9 | 1551 | 52 | |
KF615853.1 | - | 3 - 4 | 449 | 52 | |
KU178414.1 | - | 7 - 8 | 2191 | 52 |