The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001248002.1 | NP_001234931.1 | 11 - 12 | 1596 | 132 |
NM_001248003.1 | NP_001234932.1 | 10 - 11 | 1527 | 132 | |
NM_001248004.1 | NP_001234933.1 | 10 - 11 | 1494 | 132 | |
NM_001248005.1 | NP_001234934.1 | 10 - 11 | 1527 | 132 | |
NM_020183.4 | NP_064568.3 | 12 - 13 | 1638 | 132 | |
XM_006719112.3 | XP_006719175.1 | 11 - 12 | 1597 | 132 | |
XM_006719114.3 | XP_006719177.1 | 11 - 12 | 1328 | 132 | |
XM_011520766.2 | XP_011519068.1 | 12 - 13 | 1639 | 132 | |
XM_011520768.2 | XP_011519070.1 | 11 - 12 | 1547 | 132 | |
XM_011520769.2 | XP_011519071.1 | 11 - 12 | 1537 | 132 | |
XM_011520770.2 | XP_011519072.1 | 12 - 13 | 1457 | 132 | |
XM_011520771.2 | XP_011519073.1 | 12 - 13 | 1423 | 132 | |
XM_017019666.1 | XP_016875155.1 | 9 - 10 | 1368 | 132 | |
XM_017019667.1 | XP_016875156.1 | 12 - 13 | 1655 | 132 | |
XM_017019668.1 | XP_016875157.1 | 12 - 13 | 1452 | 132 | |
XM_017019669.1 | XP_016875158.1 | 11 - 12 | 1299 | 132 | |
GenBank mRNA | AB039921.1 | - | 9 - 10 | 1170 | 132 |
AF231338.1 | - | 10 - 11 | 1323 | 132 | |
AF231339.1 | - | 9 - 10 | 1284 | 132 | |
AF246960.1 | - | 12 - 13 | 1437 | 132 | |
AF246961.1 | - | 11 - 12 | 1395 | 132 | |
AF246962.1 | - | 10 - 11 | 1326 | 132 | |
AF246963.1 | - | 10 - 11 | 1293 | 132 | |
AF256215.1 | - | - | 1536 | 132 | |
AK296706.1 | - | 10 - 11 | 1461 | 132 | |
BC000172.3 | - | 9 - 10 | 1245 | 132 | |
BC125061.1 | - | 10 - 11 | 1288 | 132 | |
BC125062.1 | - | 10 - 11 | 1326 | 132 |