The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023072.2 | NP_075560.2 | 12 - 13 | 2305 | 110 |
XM_005260023.3 | XP_005260080.1 | 13 - 14 | 2467 | 110 | |
XM_017027153.1 | XP_016882642.1 | 13 - 14 | 2738 | 110 | |
XM_017027154.1 | XP_016882643.1 | 13 - 14 | 2670 | 110 | |
XM_017027155.1 | XP_016882644.1 | 12 - 13 | 2571 | 110 | |
XM_017027156.1 | XP_016882645.1 | 12 - 13 | 2242 | 110 | |
GenBank mRNA | AK022283.1 | - | 1 - 2 | 88 | 110 |
AK024452.1 | - | 12 - 13 | 2305 | 110 | |
AK299057.1 | - | 11 - 12 | 1843 | 110 | |
BG764264.1 | - | 4 - 5 | 507 | 110 |