The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143944.1 | NP_001137416.1 | 7 - 8 | 792 | 58 |
NM_181336.3 | NP_851853.1 | 8 - 9 | 1373 | 58 | |
GenBank mRNA | AK024858.1 | - | 2 - 3 | 199 | 58 |
AK074959.1 | - | 8 - 9 | 1426 | 58 | |
AK127084.1 | - | 4 - 5 | 3314 | 58 | |
AK301083.1 | - | 7 - 8 | 786 | 58 | |
AK309888.1 | - | 4 - 5 | 550 | 58 | |
BC051803.1 | - | 8 - 9 | 1363 | 58 |