The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146108.1 | NP_001139580.1 | 8 - 9 | 1023 | 65 |
NM_001146109.1 | NP_001139581.1 | 8 - 9 | 1023 | 65 | |
NM_012212.3 | NP_036344.2 | 8 - 9 | 863 | 65 | |
XM_011518395.2 | XP_011516697.1 | 5 - 6 | 699 | 65 | |
GenBank mRNA | AK091948.1 | - | 9 - 10 | 932 | 65 |
AK289597.1 | - | 8 - 9 | 1023 | 65 | |
AK298379.1 | - | 8 - 9 | 1003 | 65 | |
AK309866.1 | - | 8 - 9 | 989 | 65 | |
BC035228.2 | - | 8 - 9 | 863 | 65 | |
D49387.1 | - | 8 - 9 | 793 | 65 |