The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288607.1 | NP_001275536.1 | 11 - 12 | 2102 | 76 |
NM_001288608.1 | NP_001275537.1 | 11 - 12 | 2102 | 76 | |
NM_019032.5 | NP_061905.2 | 10 - 11 | 2033 | 76 | |
NM_025008.4 | NP_079284.2 | 10 - 11 | 2033 | 76 | |
XM_011509644.2 | XP_011507946.1 | 10 - 11 | 2064 | 76 | |
XM_011509645.2 | XP_011507947.1 | 9 - 10 | 1992 | 76 | |
XM_011509648.2 | XP_011507950.1 | 10 - 11 | 2018 | 76 | |
XM_011509649.2 | XP_011507951.1 | 10 - 11 | 2072 | 76 | |
XM_011509650.2 | XP_011507952.1 | 10 - 11 | 2072 | 76 | |
XM_011509651.2 | XP_011507953.1 | 4 - 5 | 814 | 76 | |
XM_011509652.1 | XP_011507954.1 | 4 - 5 | 520 | 76 | |
XM_017001506.1 | XP_016856995.1 | 11 - 12 | 2473 | 76 | |
GenBank mRNA | AF370411.1 | - | 4 - 5 | 569 | 76 |
AK092509.1 | - | 1 - 2 | 153 | 76 | |
AY358122.1 | - | 9 - 10 | 1847 | 76 | |
BC027478.1 | - | 3 - 4 | 472 | 76 | |
BC071852.1 | - | 2 - 3 | 329 | 76 | |
BC094811.1 | - | 2 - 3 | 329 | 76 | |
BC140800.1 | - | 10 - 11 | 1954 | 76 | |
BC144581.1 | - | 10 - 11 | 1954 | 76 |