The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324338.1 | NP_001311267.1 | 4 - 5 | 583 | 116 |
NM_152597.4 | NP_689810.3 | 4 - 5 | 680 | 116 | |
XM_011521305.2 | XP_011519607.1 | 4 - 5 | 556 | 116 | |
XM_011521306.1 | XP_011519608.1 | 4 - 5 | 969 | 116 | |
XM_011521307.2 | XP_011519609.1 | 4 - 5 | 556 | 116 | |
XM_011521309.2 | XP_011519611.1 | 4 - 5 | 556 | 116 | |
GenBank mRNA | AK093308.1 | - | 4 - 5 | 662 | 116 |
AY260140.1 | - | 3 - 4 | 477 | 116 | |
BC045191.2 | - | 4 - 5 | 672 | 116 |