Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM117B
Entrez Gene ID
150864
Gene Name
family with sequence similarity 117 member B
Gene Aliases
ALS2CR13
Chromosome Location
Chr.2: 202635178 - 202769757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173511.3 NP_775782.2 6 - 7 1334 68
GenBank mRNA AK096090.1 - 6 - 7 918 68
BC065010.1 - 5 - 6 740 68
BC106906.2 - 5 - 6 683 68
BC106907.2 - 5 - 6 679 68

View Full Product Details