Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM58
Entrez Gene ID
25893
Gene Name
tripartite motif containing 58
Gene Aliases
BIA2
Chromosome Location
Chr.1: 247857199 - 247880138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015431.3 NP_056246.3 4 - 5 813 61
GenBank mRNA AF327057.1 - 4 - 5 813 61
AK096188.1 - 4 - 5 695 61
BC074748.2 - 3 - 4 357 61
BX644877.1 - 4 - 5 369 61

View Full Product Details