Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRFN5
Entrez Gene ID
145581
Gene Name
leucine rich repeat and fibronectin type III domain containing 5
Gene Aliases
C14orf146, FIGLER8, SALM5
Chromosome Location
Chr.14: 41604684 - 41904549 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152447.3 NP_689660.2 1 - 2 1004 62
XM_005267369.3 XP_005267426.1 1 - 2 3881 62
GenBank mRNA AK055365.1 - 1 - 2 324 62
AK096627.1 - 1 - 2 977 62
AK299632.1 - 1 - 2 318 62
BC043165.1 - 1 - 2 429 62

View Full Product Details