The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080868.2 | NP_543144.1 | 1 - 2 | 535 | 118 |
XM_006710345.3 | XP_006710408.1 | 2 - 3 | 367 | 118 | |
GenBank mRNA | AF403035.1 | - | 1 - 2 | 395 | 118 |
AF429968.1 | - | 1 - 2 | 512 | 118 | |
AK098606.1 | - | 1 - 2 | 535 | 118 | |
BC036573.2 | - | 1 - 2 | 513 | 118 |