The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020890.2 | NP_065941.2 | 19 - 20 | 2678 | 117 |
XM_006713716.3 | XP_006713779.1 | 19 - 20 | 2462 | 117 | |
XM_006713717.2 | XP_006713780.1 | 20 - 21 | 2574 | 117 | |
XM_011513056.1 | XP_011511358.1 | 19 - 20 | 2422 | 117 | |
XM_011513057.2 | XP_011511359.1 | 20 - 21 | 2543 | 117 | |
GenBank mRNA | AB040957.1 | - | 19 - 20 | 2405 | 117 |
AF334474.1 | - | 19 - 20 | 2477 | 117 | |
AK022912.1 | - | 10 - 11 | 1289 | 117 | |
AK308315.1 | - | 19 - 20 | 2424 | 118 | |
AL713689.1 | - | 19 - 20 | 2473 | 117 | |
BC017659.1 | - | 5 - 6 | 551 | 117 | |
BC130564.1 | - | 19 - 20 | 2445 | 117 | |
BC136371.1 | - | 19 - 20 | 2445 | 117 |