The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252216.1 | NP_001239145.1 | 4 - 5 | 601 | 66 |
NM_213609.3 | NP_998774.2 | 4 - 5 | 875 | 66 | |
XM_017006386.1 | XP_016861875.1 | 3 - 4 | 1022 | 66 | |
XM_017006387.1 | XP_016861876.1 | 4 - 5 | 1514 | 66 | |
XM_017006388.1 | XP_016861877.1 | 4 - 5 | 1307 | 66 | |
XM_017006389.1 | XP_016861878.1 | 3 - 4 | 1019 | 66 | |
XM_017006390.1 | XP_016861879.1 | 4 - 5 | 1515 | 66 | |
GenBank mRNA | AK289668.1 | - | 4 - 5 | 864 | 66 |
AK290446.1 | - | 4 - 5 | 797 | 66 | |
AL713702.1 | - | 3 - 4 | 784 | 66 | |
AY325114.1 | - | 4 - 5 | 601 | 66 | |
BC025746.1 | - | 4 - 5 | 777 | 66 |