Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAMTA2
Entrez Gene ID
23125
Gene Name
calmodulin binding transcription activator 2
Gene Aliases
-
Chromosome Location
Chr.17: 4967992 - 4988297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171166.1 NP_001164637.1 13 - 14 2325 75
NM_001171167.1 NP_001164638.1 14 - 15 2764 75
NM_001171168.1 NP_001164639.1 13 - 14 2692 75
NM_015099.3 NP_055914.2 14 - 15 2420 75
XM_006721478.3 XP_006721541.1 14 - 15 3067 75
XM_006721481.3 XP_006721544.1 13 - 14 2980 75
XM_006721482.3 XP_006721545.1 12 - 13 2113 75
XM_011523746.2 XP_011522048.1 14 - 15 2427 75
XM_011523747.2 XP_011522049.1 14 - 15 2411 75
XM_011523748.2 XP_011522050.1 14 - 15 2426 75
XM_011523749.2 XP_011522051.1 14 - 15 2410 75
GenBank mRNA AB020716.1 - 14 - 15 2381 75
AK307947.1 - 6 - 7 756 75
AK308666.1 - 6 - 7 729 75
AL831849.1 - 13 - 14 2692 75
AL833974.1 - 13 - 14 2310 75
BC010050.2 - 6 - 7 833 75
BC016163.1 - 1 - 2 40 75
BC136534.1 - 13 - 14 2349 75
BC142695.1 - 14 - 15 2411 75
BC144232.1 - 12 - 13 2277 75
BC167148.1 - 14 - 15 2381 75
BC167149.1 - 14 - 15 2381 75
BC167151.1 - 14 - 15 2381 75
BC167160.1 - 14 - 15 2381 75

View Full Product Details