The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029939.3 | NP_001025110.2 | 5 - 6 | 899 | 81 |
NM_001167583.2 | NP_001161055.1 | 4 - 5 | 740 | 81 | |
NM_001174158.1 | NP_001167629.1 | 4 - 5 | 740 | 81 | |
NM_001289394.1 | NP_001276323.1 | 5 - 6 | 976 | 81 | |
NM_020863.3 | NP_065914.2 | 4 - 5 | 817 | 81 | |
NR_110323.1 | 4 - 5 | 817 | 81 | ||
XM_011517203.1 | XP_011515505.1 | 4 - 5 | 714 | 81 | |
XM_011517204.2 | XP_011515506.1 | 3 - 4 | 537 | 81 | |
XM_011517206.1 | XP_011515508.1 | 4 - 5 | 738 | 81 | |
XM_017013716.1 | XP_016869205.1 | 3 - 4 | 612 | 81 | |
GenBank mRNA | AB040918.1 | - | 2 - 3 | 215 | 81 |
AB167738.1 | - | 4 - 5 | 806 | 81 | |
AB167739.1 | - | 5 - 6 | 965 | 81 | |
AB167740.1 | - | 5 - 6 | 892 | 81 | |
AB167741.1 | - | 4 - 5 | 733 | 81 | |
AK300133.1 | - | 4 - 5 | 729 | 81 | |
AK300324.1 | - | 4 - 5 | 736 | 81 | |
AK300471.1 | - | 4 - 5 | 802 | 81 | |
AK311494.1 | - | 4 - 5 | 738 | 81 | |
AL832066.1 | - | 4 - 5 | 817 | 81 | |
BC012989.2 | - | 4 - 5 | 731 | 81 | |
BC025423.2 | - | 2 - 3 | 409 | 81 | |
BC098405.1 | - | 4 - 5 | 740 | 81 | |
BC101766.1 | - | 4 - 5 | 730 | 81 | |
BC101768.1 | - | 4 - 5 | 730 | 81 |