The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080850.2 | NP_001074319.1 | 11 - 12 | 1751 | 126 |
XM_011542067.2 | XP_011540369.1 | 12 - 13 | 2683 | 126 | |
XM_011542068.2 | XP_011540370.1 | 12 - 13 | 2688 | 126 | |
XM_011542069.2 | XP_011540371.1 | 11 - 12 | 2542 | 126 | |
XM_011542070.2 | XP_011540372.1 | 10 - 11 | 1709 | 126 | |
XM_011542071.2 | XP_011540373.1 | 9 - 10 | 2118 | 126 | |
XM_017002225.1 | XP_016857714.1 | 11 - 12 | 1768 | 126 | |
XM_017002226.1 | XP_016857715.1 | 12 - 13 | 2079 | 126 | |
GenBank mRNA | AL832217.1 | - | 18 - 19 | 2784 | 126 |
AY639646.1 | - | 10 - 11 | 1576 | 126 |