The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080850.2 | NP_001074319.1 | 12 - 13 | 1887 | 101 |
XM_011542067.2 | XP_011540369.1 | 13 - 14 | 2819 | 101 | |
XM_011542068.2 | XP_011540370.1 | 13 - 14 | 2824 | 101 | |
XM_011542069.2 | XP_011540371.1 | 12 - 13 | 2678 | 101 | |
XM_011542070.2 | XP_011540372.1 | 11 - 12 | 1845 | 101 | |
XM_011542071.2 | XP_011540373.1 | 10 - 11 | 2254 | 101 | |
XM_017002225.1 | XP_016857714.1 | 12 - 13 | 1904 | 101 | |
XM_017002226.1 | XP_016857715.1 | 13 - 14 | 2215 | 101 | |
GenBank mRNA | AL832217.1 | - | 19 - 20 | 2920 | 101 |
AY639646.1 | - | 11 - 12 | 1712 | 101 |