Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAUS6
Entrez Gene ID
54801
Gene Name
HAUS augmin like complex subunit 6
Gene Aliases
Dgt6, FAM29A
Chromosome Location
Chr.9: 19053137 - 19102942 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270890.1 NP_001257819.1 3 - 4 594 105
NM_017645.4 NP_060115.3 3 - 4 594 105
GenBank mRNA AF533709.1 - 3 - 4 776 105
AF537091.1 - 3 - 4 556 105
AK056458.1 - 3 - 4 556 105
AL832495.1 - 3 - 4 554 105
BC026178.1 - 3 - 4 557 105
BC065935.1 - 1 - 2 94 105
BC113981.1 - 3 - 4 391 105
BC114492.1 - 3 - 4 391 105

View Full Product Details