Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC10
Entrez Gene ID
54431
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Gene Aliases
ERdj5, JPDI, MTHr, PDIA19
Chromosome Location
Chr.2: 182716041 - 182780025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271581.1 NP_001258510.1 13 - 14 1813 122
NM_018981.2 NP_061854.1 14 - 15 1951 122
NR_073365.1 14 - 15 2705 122
NR_073366.1 14 - 15 2705 122
NR_073367.1 12 - 13 1769 122
GenBank mRNA AF490904.1 - 13 - 14 1561 122
AK027450.1 - 12 - 13 1292 122
AK027647.1 - 14 - 15 1720 122
AK027696.1 - 14 - 15 1699 122
AK027701.2 - 11 - 12 1073 122
AK074905.1 - 1 - 2 67 122
AK298138.1 - 9 - 10 798 122
AL137648.1 - 3 - 4 501 122
AL832267.1 - 7 - 8 651 122
AL832632.2 - 14 - 15 2476 122
AL832646.3 - 12 - 13 1302 122
AY089971.1 - 10 - 11 1123 122
AY358577.1 - 13 - 14 1570 122
BC070034.1 - 14 - 15 2445 122
BC117299.1 - 14 - 15 1666 122
BC126168.1 - 14 - 15 1666 122

View Full Product Details