Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJC10
Entrez Gene ID
54431
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Gene Aliases
ERdj5, JPDI, MTHr, PDIA19
Chromosome Location
Chr.2: 182716041 - 182780025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271581.1 NP_001258510.1 19 - 20 2500 91
NM_018981.2 NP_061854.1 20 - 21 2638 91
NR_073365.1 20 - 21 3392 91
NR_073366.1 21 - 22 3495 91
NR_073367.1 19 - 20 2531 91
GenBank mRNA AF038503.1 - 20 - 21 2349 91
AF314529.1 - 7 - 8 623 91
AF490904.1 - 19 - 20 2248 91
AK027450.1 - 18 - 19 1979 91
AK027647.1 - 20 - 21 2407 91
AK027696.1 - 20 - 21 2386 91
AK027701.2 - 17 - 18 1760 91
AK074905.1 - 7 - 8 754 91
AK298138.1 - 15 - 16 1485 91
AL137648.1 - 9 - 10 1188 91
AL832267.1 - 13 - 14 1338 91
AL832632.2 - 20 - 21 3163 91
AL832646.3 - 18 - 19 1989 91
AY089971.1 - 17 - 18 1885 91
AY358577.1 - 19 - 20 2257 91
BC070034.1 - 21 - 22 3235 91
BC117299.1 - 20 - 21 2353 91
BC126168.1 - 20 - 21 2353 91

View Full Product Details