The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010854.1 | NP_001010854.1 | 2 - 3 | 397 | 86 |
XM_017021044.1 | XP_016876533.1 | 2 - 3 | 361 | 86 | |
XM_017021045.1 | XP_016876534.1 | 2 - 3 | 361 | 86 | |
XM_017021047.1 | XP_016876536.1 | 2 - 3 | 194 | 86 | |
GenBank mRNA | AL832848.1 | - | 1 - 2 | 75 | 86 |
BG489583.1 | - | 1 - 2 | 63 | 85 | |
CB988671.1 | - | - | 424 | 86 |