Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC7A
Entrez Gene ID
57217
Gene Name
tetratricopeptide repeat domain 7A
Gene Aliases
GIDID, MINAT, TTC7
Chromosome Location
Chr.2: 46915931 - 47076136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288951.1 NP_001275880.1 12 - 13 1878 79
NM_001288953.1 NP_001275882.1 13 - 14 1869 79
NM_001288955.1 NP_001275884.1 11 - 12 1720 79
NM_020458.3 NP_065191.2 12 - 13 1878 79
XM_005264439.3 XP_005264496.1 13 - 14 2031 79
XM_011532998.2 XP_011531300.1 14 - 15 2096 79
XM_011532999.2 XP_011531301.1 12 - 13 1821 79
XM_011533000.2 XP_011531302.1 9 - 10 953 79
XM_011533001.2 XP_011531303.1 5 - 6 743 79
XM_017004524.1 XP_016860013.1 12 - 13 1822 79
XM_017004525.1 XP_016860014.1 12 - 13 1363 79
XM_017004527.1 XP_016860016.1 12 - 13 1973 79
XM_017004528.1 XP_016860017.1 13 - 14 2038 79
XM_017004529.1 XP_016860018.1 12 - 13 1821 79
XM_017004530.1 XP_016860019.1 12 - 13 1822 79
GenBank mRNA AB032966.2 - 13 - 14 1458 79
AK056464.1 - 13 - 14 1841 79
AL834383.1 - 12 - 13 1878 79
BC001978.1 - 3 - 4 287 79
BC065554.1 - 9 - 10 996 79
BC111487.1 - 12 - 13 1766 79
BC114365.2 - 11 - 12 1351 79

View Full Product Details