Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2F7
Entrez Gene ID
144455
Gene Name
E2F transcription factor 7
Gene Aliases
-
Chromosome Location
Chr.12: 77021246 - 77065580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203394.2 NP_976328.2 4 - 5 773 104
XM_011537966.2 XP_011536268.1 4 - 5 773 104
XM_011537969.2 XP_011536271.1 3 - 4 336 104
GenBank mRNA AK097677.1 - 2 - 3 366 104
BC136366.1 - 4 - 5 612 104
BC136367.1 - 4 - 5 612 104

View Full Product Details