The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153451.2 | NP_703152.1 | 2 - 3 | 316 | 62 |
XM_006718470.2 | XP_006718533.1 | 2 - 3 | 273 | 62 | |
GenBank mRNA | AF503940.1 | - | 2 - 3 | 266 | 62 |
AK026600.1 | - | 2 - 3 | 335 | 62 | |
AK294004.1 | - | 2 - 3 | 264 | 62 | |
AK311918.1 | - | 2 - 3 | 266 | 62 | |
BC006448.2 | - | 2 - 3 | 232 | 62 | |
BC019024.1 | - | 2 - 3 | 233 | 62 | |
BC065542.1 | - | 2 - 3 | 259 | 62 |