The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183376.2 | NP_899232.2 | 1 - 2 | 468 | 123 |
GenBank mRNA | AK290221.1 | - | 1 - 2 | 468 | 123 |
BC028704.2 | - | 1 - 2 | 458 | 123 | |
BC070100.1 | - | 1 - 2 | 448 | 123 | |
DB089068.1 | - | 1 - 2 | 473 | 123 |