Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENKD1
Entrez Gene ID
84080
Gene Name
enkurin domain containing 1
Gene Aliases
C16orf48, DAKV6410
Chromosome Location
Chr.16: 67662947 - 67666725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032140.1 NP_115516.1 4 - 5 864 101
GenBank mRNA AL136786.1 - 4 - 5 864 101
AY358834.1 - 1 - 2 578 101
BC008284.1 - 4 - 5 831 101

View Full Product Details