The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320925.1 | NP_001307854.1 | 2 - 3 | 696 | 105 |
NM_032356.4 | NP_115732.2 | 1 - 2 | 880 | 105 | |
XM_017025226.1 | XP_016880715.1 | 4 - 5 | 465 | 105 | |
GenBank mRNA | AF530060.1 | - | 2 - 3 | 268 | 105 |
BC006407.2 | - | 2 - 3 | 367 | 105 | |
BC033861.1 | - | 1 - 2 | 843 | 105 | |
BC051846.1 | - | 2 - 3 | 696 | 105 | |
BC059944.1 | - | 2 - 3 | 296 | 105 |