The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321335.1 | NP_001308264.1 | 2 - 3 | 614 | 140 |
NM_170746.3 | NP_734467.1 | 2 - 3 | 614 | 140 | |
GenBank mRNA | AF536829.1 | - | 2 - 3 | 510 | 140 |
BC021122.1 | - | 2 - 3 | 328 | 140 | |
BG714263.1 | - | 2 - 3 | 329 | 140 |