The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127361.2 | NP_001120833.1 | 6 - 7 | 1528 | 105 |
NM_001300826.1 | NP_001287755.1 | 6 - 7 | 1528 | 105 | |
NM_153341.3 | NP_699172.2 | 6 - 7 | 1531 | 105 | |
XM_006710356.2 | XP_006710419.1 | 6 - 7 | 1984 | 105 | |
XM_006710357.3 | XP_006710420.1 | 6 - 7 | 1984 | 105 | |
XM_006710358.3 | XP_006710421.1 | 6 - 7 | 1984 | 105 | |
GenBank mRNA | AK074486.1 | - | 6 - 7 | 831 | 105 |
AK131439.1 | - | 7 - 8 | 1225 | 105 | |
BC020595.2 | - | 2 - 3 | 236 | 105 | |
BC062374.1 | - | 6 - 7 | 1005 | 105 | |
BC113560.1 | - | 6 - 7 | 898 | 105 | |
BC143687.1 | - | 6 - 7 | 895 | 105 |