Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD2
Entrez Gene ID
143162
Gene Name
FERM and PDZ domain containing 2
Gene Aliases
PDZD5C, PDZK4, PDZK5C
Chromosome Location
Chr.10: 48156559 - 48274898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018071.3 NP_001018081.3 24 - 25 3410 128
NM_001042512.2 NP_001035977.2 1 - 2 278 128
NM_001318191.1 NP_001305120.1 22 - 23 3051 128
GenBank mRNA AK123038.1 - 22 - 23 3051 128
AY491519.1 - 1 - 2 277 128
BC031614.1 - 3 - 4 273 128
BC073954.1 - 1 - 2 259 128
BC144636.1 - 1 - 2 219 128
BC144639.1 - 1 - 2 219 128
CR749241.1 - 24 - 25 3382 128

Gene Symbol
FRMPD2B
Entrez Gene ID
728798
Gene Name
FERM and PDZ domain containing 2B, pseudogene
Gene Aliases
FRMPD2L1, FRMPD2L2, FRMPD2P1, FRMPD2P2, FRMPDP2, PDZD5A, PDZD5B, PDZK5A, PDZK5B, bA556L1.2, yX59F3.2
Chromosome Location
Chr.10: 46876674 - 46895326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033172.1 1 - 2 332 128
GenBank mRNA AK294650.1 - 1 - 2 287 128
AK313659.1 - 1 - 2 289 128
AK315971.1 - 1 - 2 332 128
BC071635.1 - 1 - 2 244 128
BC072395.1 - 1 - 2 258 128
BC110798.1 - 1 - 2 255 128
BC137431.1 - 1 - 2 219 128
BC137434.1 - 1 - 2 219 128
BC137439.1 - 1 - 2 217 128
BC137440.1 - 1 - 2 219 128
BC144635.1 - 1 - 2 219 128
DC315823.1 - 1 - 2 332 128

View Full Product Details