Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZC2HC1B
- Entrez Gene ID
-
153918
- Gene Name
- zinc finger C2HC-type containing 1B
- Gene Aliases
-
C6orf94, FAM164B, dJ468K18.5
- Chromosome Location
-
Chr.6: 143864436 - 143938346 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC171862.1 |
- |
2 - 3 |
220 |
73 |
View Full Product Details