Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL29
Entrez Gene ID
114818
Gene Name
kelch like family member 29
Gene Aliases
KBTBD9
Chromosome Location
Chr.2: 23385419 - 23708613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052920.1 NP_443152.1 1 - 2 357 127

View Full Product Details