Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF27
Entrez Gene ID
55582
Gene Name
kinesin family member 27
Gene Aliases
-
Chromosome Location
Chr.9: 83832795 - 83921539 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271927.1 NP_001258856.1 11 - 12 2739 99
NM_017576.2 NP_060046.1 12 - 13 2937 99
XM_011518848.2 XP_011517150.1 13 - 14 3016 99
XM_011518849.2 XP_011517151.1 14 - 15 3132 99
XM_011518850.2 XP_011517152.1 13 - 14 3105 99
XM_011518856.2 XP_011517158.1 14 - 15 3235 99
XM_011518857.2 XP_011517159.1 14 - 15 2701 99
XM_011518858.2 XP_011517160.1 10 - 11 1489 99
XM_017014900.1 XP_016870389.1 15 - 16 3284 99
XM_017014901.1 XP_016870390.1 15 - 16 3166 99
XM_017014902.1 XP_016870391.1 14 - 15 3212 99
XM_017014903.1 XP_016870392.1 13 - 14 3157 99
XM_017014904.1 XP_016870393.1 13 - 14 3054 99
XM_017014905.1 XP_016870394.1 13 - 14 3037 99
XM_017014906.1 XP_016870395.1 14 - 15 3235 99
XM_017014907.1 XP_016870396.1 12 - 13 2959 99
XM_017014909.1 XP_016870398.1 13 - 14 2837 99
XM_017014911.1 XP_016870400.1 11 - 12 2331 99
XM_017014912.1 XP_016870401.1 11 - 12 1976 99
XM_017014913.1 XP_016870402.1 12 - 13 2168 99
XM_017014915.1 XP_016870404.1 12 - 13 2138 99
XM_017014916.1 XP_016870405.1 13 - 14 2743 99
GenBank mRNA AL133654.1 - 7 - 8 1052 99
AY237536.1 - 12 - 13 2899 99
AY237537.1 - 11 - 12 2701 99
BC140788.1 - 12 - 13 2898 99

View Full Product Details