Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MROH2A
Entrez Gene ID
339766
Gene Name
maestro heat like repeat family member 2A
Gene Aliases
HEATR7B1
Chromosome Location
Chr.2: 233774022 - 233833423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287395.1 NP_001274324.1 40 - 41 4975 105
XM_011511075.2 XP_011509377.1 40 - 41 5182 105
XM_011511076.2 XP_011509378.1 40 - 41 5170 105
XM_011511077.2 XP_011509379.1 40 - 41 5152 105
XM_011511078.2 XP_011509380.1 40 - 41 5143 105
XM_011511079.2 XP_011509381.1 40 - 41 5137 105
XM_011511080.2 XP_011509382.1 40 - 41 6653 105
XM_011511081.2 XP_011509383.1 39 - 40 5077 105
XM_011511082.2 XP_011509384.1 39 - 40 5044 105
XM_011511084.2 XP_011509386.1 39 - 40 4984 105
XM_011511085.2 XP_011509387.1 34 - 35 4252 105
XM_011511086.2 XP_011509388.1 31 - 32 3907 105
GenBank mRNA BC042133.1 - 5 - 6 802 105

View Full Product Details