Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTSL4
Entrez Gene ID
54507
Gene Name
ADAMTS like 4
Gene Aliases
ADAMTSL-4, ECTOL2, TSRC1
Chromosome Location
Chr.1: 150549369 - 150560937 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288607.1 NP_001275536.1 5 - 6 729 71
NM_001288608.1 NP_001275537.1 5 - 6 729 71
NM_019032.5 NP_061905.2 5 - 6 729 71
NM_025008.4 NP_079284.2 5 - 6 729 71
XM_011509644.2 XP_011507946.1 4 - 5 691 71
XM_011509645.2 XP_011507947.1 4 - 5 688 71
XM_011509648.2 XP_011507950.1 4 - 5 645 71
XM_011509649.2 XP_011507951.1 4 - 5 699 71
XM_011509650.2 XP_011507952.1 4 - 5 699 71
XM_017001506.1 XP_016856995.1 5 - 6 1100 71
GenBank mRNA AL513944.3 - 5 - 6 675 71
AY358122.1 - 4 - 5 543 71
BC140800.1 - 4 - 5 581 71
BC144581.1 - 4 - 5 581 71
BG005397.1 - 1 - 2 140 71

View Full Product Details