The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015243.2 | NP_056058.2 | 12 - 13 | 1764 | 137 |
NM_017890.4 | NP_060360.3 | 12 - 13 | 1764 | 137 | |
NM_152564.4 | NP_689777.3 | 12 - 13 | 1764 | 137 | |
XM_005250800.3 | XP_005250857.1 | 12 - 13 | 2070 | 137 | |
XM_005250801.4 | XP_005250858.1 | 12 - 13 | 7940 | 137 | |
XM_006716510.3 | XP_006716573.1 | 12 - 13 | 1756 | 137 | |
XM_011516848.2 | XP_011515150.1 | 12 - 13 | 1756 | 137 | |
XM_011516849.2 | XP_011515151.1 | 12 - 13 | 1756 | 137 | |
XM_011516850.2 | XP_011515152.1 | 10 - 11 | 1396 | 137 | |
XM_011516853.2 | XP_011515155.1 | 12 - 13 | 1756 | 137 | |
XM_011516859.2 | XP_011515161.1 | 12 - 13 | 1756 | 137 | |
XM_011516861.1 | XP_011515163.1 | 12 - 13 | 1756 | 137 | |
XM_011516865.1 | XP_011515167.1 | 12 - 13 | 1756 | 137 | |
XM_011516866.2 | XP_011515168.1 | 12 - 13 | 1756 | 137 | |
XM_017013109.1 | XP_016868598.1 | 12 - 13 | 1756 | 137 | |
XM_017013110.1 | XP_016868599.1 | 5 - 6 | 554 | 137 | |
GenBank mRNA | AJ608772.1 | - | 12 - 13 | 1756 | 137 |
AJ608773.1 | - | 12 - 13 | 1756 | 137 | |
AK091431.1 | - | 1 - 2 | 104 | 137 | |
AY223814.1 | - | 12 - 13 | 1764 | 137 | |
AY223815.1 | - | 12 - 13 | 1764 | 137 | |
AY223816.1 | - | 12 - 13 | 1764 | 137 | |
AY223817.1 | - | 12 - 13 | 1764 | 137 | |
BX098890.1 | - | 2 - 3 | 150 | 137 |