The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198471.2 | NP_940873.2 | 5 - 6 | 1700 | 58 |
XM_006722718.3 | XP_006722781.1 | 5 - 6 | 1819 | 58 | |
XM_011527884.2 | XP_011526186.1 | 5 - 6 | 2076 | 58 | |
XM_017026563.1 | XP_016882052.1 | 4 - 5 | 1836 | 58 | |
GenBank mRNA | AB284126.1 | - | 5 - 6 | 1700 | 58 |
AK128815.1 | - | 5 - 6 | 1700 | 58 | |
BC066124.1 | - | 5 - 6 | 1700 | 58 | |
BC066775.1 | - | 5 - 6 | 1700 | 58 |